Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053043.2 | NP_444271.2 | 18 | 4927 | ||
XM_005249531.1 | XP_005249588.1 | 19 | 5095 | |||
XM_005249532.1 | XP_005249589.1 | 19 | 5092 | |||
XM_005249533.1 | XP_005249590.1 | 19 | 5065 | |||
XM_005249534.1 | XP_005249591.1 | 18 | 4975 | |||
XM_005249535.1 | XP_005249592.1 | 18 | 4945 | |||
XM_011515840.1 | XP_011514142.1 | 18 | 4885 | |||
GenBank mRNA | AK055645.1 | - | 1 | 539 | ||
AK056415.1 | - | 3 | 1335 | |||
AL832196.1 | - | 8 | 1661 |