Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008420.2 | NP_001008420.1 | 6 | 1087 | ||
GenBank mRNA | BC086655.1 | - | 6 | 1087 | 30542783 |