Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008496.3 | NP_001008496.2 | 15, 16 | 2361 | ||
NM_001255975.1 | NP_001242904.1 | 15, 16 | 2334 | |||
NR_045648.1 | 16, 17 | 2544 | ||||
NR_045649.1 | 16, 17 | 2417 | ||||
GenBank mRNA | AB079368.1 | - | 15, 16 | 2357 | ||
AK302409.1 | - | 15, 16 | 2334 | |||
AK315806.1 | - | 15, 16 | 2334 | |||
BC144693.1 | - | 16, 17 | 2174 | |||
BC144696.1 | - | 16, 17 | 2301 |