Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008785.5 | NP_001008785.2 | 4 | 3167 | ||
NM_001102670.2 | NP_001096140.1 | 3 | 2956 | |||
GenBank mRNA | AB093302.1 | - | 2 | 2081 | ||
AK136095.1 | - | 1 | 274 | |||
AK142080.1 | - | 4 | 3141 | |||
AK167507.1 | - | 3 | 2907 | |||
BC088734.1 | - | 3 | 3076 | 30459462 |