Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008791.2 | NP_001008791.1 | 7 | 2316 | ||
NM_001008792.2 | NP_001008792.1 | 7 | 2328 | |||
NM_001008793.2 | NP_001008793.1 | 7 | 2316 | |||
NM_028640.2 | NP_082916.1 | 7 | 2316 | |||
GenBank mRNA | AK021254.1 | - | 1 | 82 | ||
AK122523.1 | - | 7 | 1447 | |||
AK157955.1 | - | 7 | 2317 | |||
AY739114.1 | - | 7 | 2276 | |||
AY739115.1 | - | 7 | 2276 | |||
AY739116.1 | - | 7 | 2288 | |||
AY739117.1 | - | 7 | 2276 | |||
AY739118.1 | - | 3 | 892 | |||
AY739119.1 | - | 3 | 892 | |||
AY739120.1 | - | 2 | 679 | |||
AY739121.1 | - | 2 | 797 | |||
AY739122.1 | - | 2 | 797 | |||
BC150757.1 | - | 7 | 1673 | 9087668 | ||
HQ148552.1 | - | 7 | 1558 |