Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009353.1 | NP_001009353.1 | 2, 3 | 329 | ||
XM_006244606.2 | XP_006244668.1 | 2, 3 | 446 | |||
GenBank mRNA | BC088457.1 | - | 2, 3 | 329 | 7127300 |