Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290225.1 | NP_001277154.1 | 1, 2 | 342 | ||
NM_014754.2 | NP_055569.1 | 1, 2 | 342 | |||
GenBank mRNA | AK293036.1 | - | 1, 2 | 326 | ||
BC002376.2 | - | 1, 2 | 315 | 2961075 | ||
BC004192.2 | - | 1, 2 | 292 | 3509418 | ||
BC004390.1 | - | 1, 2 | 292 | 3634879 | ||
D14694.1 | - | 1, 2 | 289 | |||
HY087183.1 | - | 1, 2 | 342 |