Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009486.1 | NP_001009486.1 | 8, 9 | 989 | ||
XM_008763404.1 | XP_008761626.1 | 7, 8 | 966 | |||
GenBank mRNA | AY649835.1 | - | 8, 9 | 989 | ||
BC085917.1 | - | 952 | 7108565 |