Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam115e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009534.1 | NP_001009534.1 | 3 | 1113 | ||
GenBank mRNA | BC061995.1 | - | 3 | 1113 | 5599703 |