Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001802.1 | NP_001793.1 | 3 | 520 | ||
GenBank mRNA | AK292273.1 | - | 3 | 267 | ||
BC017503.1 | - | 3 | 502 | 4873337 | ||
BG386156.1 | - | 580 | ||||
BG750955.1 | - | 3 | 503 | |||
D12981.1 | - | 3 | 196 | |||
M63256.1 | - | 3 | 380 |