Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 22 | 3133 | ||
NM_001278470.1 | NP_001265399.1 | 22 | 3133 | |||
NM_001278471.1 | NP_001265400.1 | 22 | 3064 | |||
NM_001278472.1 | NP_001265401.1 | 23 | 3211 | |||
NM_015026.2 | NP_055841.2 | 22 | 3133 | |||
XM_017019041.1 | XP_016874530.1 | 23 | 3136 | |||
XM_017019042.1 | XP_016874531.1 | 23 | 3136 | |||
XM_017019043.1 | XP_016874532.1 | 24 | 3210 | |||
XM_017019044.1 | XP_016874533.1 | 24 | 3212 | |||
XM_017019045.1 | XP_016874534.1 | 25 | 2880 | |||
XM_017019046.1 | XP_016874535.1 | 19 | 2305 | |||
GenBank mRNA | AB017814.1 | - | 22 | 2858 | ||
AB028963.3 | - | 23 | 3104 | |||
AL833066.2 | - | 22 | 3128 | |||
AL834320.1 | - | 16 | 2091 | |||
BC136621.1 | - | 22 | 3102 | 9020611 | ||
BC141817.1 | - | 23 | 3104 | 40148050 | ||
BC142710.1 | - | 23 | 3104 | 40148826 | ||
BC144309.1 | - | 22 | 3033 | 9052829 | ||
BC151241.1 | - | 23 | 3127 | 8860514 | ||
BX537415.1 | - | 22 | 3131 |