Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015130.2 | NP_055945.2 | 12 | 2257 | ||
GenBank mRNA | AB020689.2 | - | 12 | 2060 | ||
AB449892.1 | - | 12 | 1985 | |||
AK127020.1 | - | 9 | 1545 | |||
BC038804.1 | - | 3 | 371 | |||
BC146758.1 | - | 12 | 2060 | 9007196 |