Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 4 | 631 | ||
NM_015291.3 | NP_056106.1 | 5 | 816 | |||
NR_109898.1 | 5 | 816 | ||||
GenBank mRNA | AB023179.2 | - | 5 | 772 | ||
AK091848.1 | - | 5 | 774 | |||
AK292479.1 | - | 5 | 748 | |||
AL833299.1 | - | 5 | 755 | |||
BC047363.1 | - | 5 | 816 | 5269539 | ||
BX648982.1 | - | 5 | 741 | |||
CR749259.1 | - | 4 | 576 | |||
CR749567.1 | - | 4 | 576 |