Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 14 | 1759 | ||
NM_020729.2 | NP_065780.2 | 13 | 1679 | |||
XM_005271054.3 | XP_005271111.1 | 13 | 1677 | |||
XM_011541815.2 | XP_011540117.1 | 14 | 1764 | |||
XM_011541819.2 | XP_011540121.1 | 14 | 1764 | |||
XM_011541823.2 | XP_011540125.1 | 14 | 1764 | |||
XM_011541824.2 | XP_011540126.1 | 14 | 1764 | |||
XM_017001876.1 | XP_016857365.1 | 14 | 1556 | |||
XM_017001877.1 | XP_016857366.1 | 13 | 1469 | |||
XM_017001878.1 | XP_016857367.1 | 13 | 1647 | |||
XM_017001879.1 | XP_016857368.1 | 13 | 1677 | |||
XM_017001880.1 | XP_016857369.1 | 12 | 1560 | |||
XM_017001881.1 | XP_016857370.1 | 11 | 1432 | |||
XM_017001882.1 | XP_016857371.1 | 7 | 1877 | |||
GenBank mRNA | AB033055.1 | - | 14 | 1679 | ||
AK302792.1 | - | 13 | 1651 | |||
BC047368.1 | - | 13 | 1619 | 5276959 | ||
BQ894001.1 | - | 426 |