Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020768.3 | NP_065819.1 | 4 | 4738 | ||
XM_005268492.2 | XP_005268549.1 | 3 | 5129 | |||
XM_005268493.2 | XP_005268550.1 | 3 | 4525 | |||
XM_006714789.2 | XP_006714852.1 | 3 | 5189 | |||
XM_011537671.2 | XP_011535973.1 | 5 | 5024 | |||
GenBank mRNA | AB037738.1 | - | 3 | 5201 | ||
CR749266.1 | - | 4 | 4738 |