Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 19 | 4201 | ||
XM_011537044.2 | XP_011535346.1 | 20 | 4303 | |||
XM_011537045.2 | XP_011535347.1 | 19 | 4113 | |||
GenBank mRNA | AL833497.1 | - | 6 | 3425 | ||
BX647116.1 | - | 19 | 4153 | |||
L40392.1 | - | 5 | 2287 |