Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021287.1 | NP_067262.1 | 24, 25 | 5437 | ||
GenBank mRNA | AK147671.1 | - | 24, 25 | 5453 | ||
AK172931.1 | - | 14, 15 | 3746 | |||
BC048851.1 | - | 2, 3 | 255 | |||
BC079628.1 | - | 2, 3 | 389 | |||
BC079860.1 | - | 24, 25 | 5437 | 6834745 |