Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021287.1 | NP_067262.1 | 37 | 7580 | ||
GenBank mRNA | AF026489.1 | - | 1 | 17 | ||
AK147671.1 | - | 37 | 7596 | |||
AK172931.1 | - | 27 | 5889 | |||
BC033305.1 | - | 3 | 379 | |||
BC048851.1 | - | 15 | 2398 | |||
BC079628.1 | - | 15 | 2532 | |||
BC079860.1 | - | 37 | 7580 | 6834745 |