Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007154.3 | NP_001007155.1 | 8 | 1305 | ||
NM_001177789.1 | NP_001171260.1 | 8 | 1523 | |||
NM_001177790.1 | NP_001171261.1 | 8 | 1523 | |||
NM_001177791.1 | NP_001171262.1 | 8 | 1357 | |||
NM_028806.2 | NP_083082.1 | 8 | 1762 | |||
GenBank mRNA | AJ632220.1 | - | 8 | 1320 | ||
AJ632221.1 | - | 8 | 1358 | |||
AJ632222.1 | - | 8 | 1305 | |||
AJ632223.1 | - | 7 | 1195 | |||
AK005136.1 | - | 8 | 1327 | |||
AK032242.1 | - | 8 | 1529 | |||
AK038449.1 | - | 8 | 1273 | |||
AK039213.1 | - | 8 | 1312 | |||
AK135144.1 | - | 8 | 1763 | |||
AK220554.1 | - | 8 | 1523 | |||
AY501387.1 | - | 8 | 1469 | |||
BC058621.1 | - | 8 | 1678 | 5708186 | ||
BC059869.1 | - | 8 | 1541 | 6809310 |