Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANC1 SIRNA Assays ›
Gene Symbol:
TANC1
Gene Name:
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases:
ROLSB, TANC
Chromosome Location:
Chr.2: 158968600 - 159232659 on Build GRCh38
UniGene ID:
Hs.61590
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219692
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145909.1
NP_001139381.1
7
860
NM_033394.2
NP_203752.2
7
863
XM_005246918.4
XP_005246975.1
6
627
XM_006712810.3
XP_006712873.1
6
750
XM_006712812.3
XP_006712875.1
6
751
XM_011512048.2
XP_011510350.1
9
1041
XM_011512051.2
XP_011510353.1
8
906
XM_011512053.2
XP_011510355.1
7
796
XM_011512055.2
XP_011510357.1
7
853
XM_011512059.2
XP_011510361.1
3
399
XM_017005141.1
XP_016860630.1
6
655
XM_017005142.1
XP_016860631.1
8
1260
XM_017005143.1
XP_016860632.1
6
945
XM_017005144.1
XP_016860633.1
6
655
XM_017005145.1
XP_016860634.1
9
1075
XM_017005146.1
XP_016860635.1
8
907
XM_017005147.1
XP_016860636.1
7
856
XM_017005148.1
XP_016860637.1
8
1022
XM_017005149.1
XP_016860638.1
5
553
XM_017005150.1
XP_016860639.1
6
658
XM_017005151.1
XP_016860640.1
6
658
XM_017005152.1
XP_016860641.1
6
655
XM_017005154.1
XP_016860643.1
8
1021
XM_017005155.1
XP_016860644.1
6
751
XM_017005158.1
XP_016860647.1
9
1004
GenBank mRNA
AK023422.1
-
2
1827
AK123161.1
-
2
1422
AK128859.1
-
7
857
BC150597.1
-
6
701
9056967
BX648327.1
-
2
123
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001002854, XM_008761838, XM_008761839, XM_006234205, XM_006234204, and others.
Gene Ontology Categories:
Function(s)
myoblast fusion
visual learning
dendritic spine maintenance
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0