Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 2279 | ||
NM_001128934.2 | NP_001122406.1 | 4 | 2279 | |||
NM_001286754.1 | NP_001273683.1 | 4 | 2231 | |||
NM_133477.2 | NP_597734.2 | 4 | 2279 | |||
GenBank mRNA | AJ010482.1 | - | 2 | 1299 | ||
AK296962.1 | - | 2 | 1045 | |||
AK307711.1 | - | 4 | 2275 | |||
AL832031.1 | - | 4 | 2262 | |||
AL832363.1 | - | 4 | 2173 | |||
AL833294.1 | - | 4 | 2265 | |||
AL833547.1 | - | 4 | 2279 | |||
BC136256.1 | - | 4 | 2154 | 9020243 | ||
BC150629.1 | - | 4 | 2278 | 9056999 | ||
BX647849.1 | - | 2 | 1788 | |||
BX648090.1 | - | 3 | 1937 | |||
CR936673.1 | - | 4 | 2271 | |||
EU481975.1 | - | 4 | 2083 | |||
FN422000.1 | - | 2 | 1334 | |||
KF147165.1 | - | 4 | 2231 |