Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001980.2 | NP_001001980.2 | 9, 10 | 1052 | ||
NM_001256122.1 | NP_001243051.1 | 4, 5 | 814 | |||
GenBank mRNA | AK019457.1 | - | 4, 5 | 671 | ||
AK122443.1 | - | 2, 3 | 167 | |||
AK134095.1 | - | 4, 5 | 615 | |||
AK147547.1 | - | 4, 5 | 815 | |||
BC042189.1 | - | 1, 2 | 131 | |||
CD804084.1 | - | 4, 5 | 512 |