Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002764.1 | NP_001002764.1 | 13, 14 | 3396 | ||
GenBank mRNA | AK129203.1 | - | 12, 13 | 2015 | ||
BC037179.1 | - | 11, 12 | 1686 | |||
BC047279.1 | - | 13, 14 | 3408 | |||
BC066040.1 | - | 13, 14 | 3396 | 6410245 | ||
BC077715.1 | - | 13, 14 | 3359 |