Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003917.4 | NP_001003917.2 | 8 | 1089 | ||
NM_001288612.1 | NP_001275541.1 | 8 | 1092 | |||
NM_001288613.1 | NP_001275542.1 | 8 | 1337 | |||
GenBank mRNA | AK210178.1 | - | 1 | 42 | ||
AM085510.1 | - | 8 | 1002 | |||
BC056488.1 | - | 8 | 1250 | |||
BC079884.1 | - | 8 | 1037 | 6840050 | ||
BC082307.1 | - | 6 | 795 |