Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003949.3 | NP_001003949.2 | 2 | 501 | ||
GenBank mRNA | AY096036.1 | - | 2 | 211 | ||
AY096037.1 | - | 2 | 211 | |||
BB645384.1 | - | 2 | 501 | |||
BC005494.1 | - | 1 | 24 | |||
BC052787.1 | - | 1 | 43 |