Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004148.4 | NP_001004148.1 | 2 | 157 | ||
GenBank mRNA | AK043499.1 | - | 2 | 158 | ||
AK048662.1 | - | 2 | 158 | |||
AK048944.1 | - | 3 | 415 | |||
AK140943.1 | - | 2 | 158 | |||
AK158253.1 | - | 2 | 158 | |||
AY368893.1 | - | 2 | 118 |