Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004238.1 | NP_001004238.1 | 7 | 977 | ||
GenBank mRNA | BC079323.1 | - | 7 | 977 | 7135453 | |
FQ226410.1 | - | 6 | 579 |