Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134657.1 | NP_001128129.1 | 1 | 1525 | ||
GenBank mRNA | AK128089.1 | - | 1 | 1522 |