Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122104.1 | 1 | 927 | |||
GenBank mRNA | AK124832.1 | - | 1 | 927 | ||
BC132910.1 | - | 1 | 643 |