Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243765.1 | NP_001230694.1 | 1 | 527 | ||
XM_017002995.1 | XP_016858484.1 | 1 | 1399 | |||
GenBank mRNA | AB007940.1 | - | 2 | 503 |