Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005343.2 | NP_001005343.1 | 2 | 1874 | ||
GenBank mRNA | AK135123.1 | - | 2 | 1873 | ||
AY591908.1 | - | 2 | 1886 | |||
BC139398.1 | - | 2 | 1798 | 8862420 | ||
BC139407.1 | - | 2 | 1798 | 8862429 |