Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005404.3 | NP_001005404.1 | 5 | 4404 | ||
XM_017024621.1 | XP_016880110.1 | 5 | 4422 | |||
GenBank mRNA | AB098736.1 | - | 5 | 4327 |