Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005506.3 | NP_001005506.2 | 10 | 4270 | ||
NM_001199695.1 | NP_001186624.1 | 11 | 3987 | |||
GenBank mRNA | AK011182.1 | - | 1 | 116 | ||
AK150240.1 | - | 11 | 3944 | |||
BC060227.1 | - | 10 | 4105 | 30357541 | ||
BC065115.1 | - | 9 | 4341 | 30362350 |