Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005507.2 | NP_001005507.1 | 14 | 1770 | ||
NM_001160256.1 | NP_001153728.1 | 14 | 1770 | |||
NM_001160257.1 | NP_001153729.1 | 14 | 1760 | |||
GenBank mRNA | BC082789.1 | - | 14 | 1757 | 30105222 | |
BC086651.1 | - | 14 | 1770 | 6851591 |