Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005540.1 | NP_001005540.1 | 10 | 1562 | ||
XM_008761380.1 | XP_008759602.1 | 11 | 2060 | |||
XM_008761381.1 | XP_008759603.1 | 10 | 1952 | |||
GenBank mRNA | BC082053.1 | - | 10 | 1562 | 7132147 |