Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM13A SIRNA Assays ›
Gene Symbol:
FAM13A
Gene Name:
family with sequence similarity 13 member A
Gene Aliases:
ARHGAP48, FAM13A1
Chromosome Location:
Chr.4: 88725954 - 89111398 on Build GRCh38
UniGene ID:
Hs.714536
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22106
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM13A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001015045.2
NP_001015045.1
4
543
NM_001265578.1
NP_001252507.1
3
501
NM_001265579.1
NP_001252508.1
4
543
NM_001265580.1
NP_001252509.1
2
459
NM_014883.3
NP_055698.2
10
1395
XM_005262681.2
XP_005262738.1
9
1353
XM_005262682.2
XP_005262739.1
10
1494
XM_005262683.2
XP_005262740.1
10
1395
XM_005262684.3
XP_005262741.1
9
1174
XM_006714057.3
XP_006714120.1
7
670
XM_011531516.1
XP_011529818.1
11
1416
XM_011531517.1
XP_011529819.1
9
1311
XM_011531518.1
XP_011529820.1
11
1545
XM_011531519.2
XP_011529821.1
9
1104
XM_017007624.1
XP_016863113.1
9
1353
XM_017007625.1
XP_016863114.1
9
1014
XM_017007626.1
XP_016863115.1
7
648
XM_017007627.1
XP_016863116.1
9
1239
XM_017007628.1
XP_016863117.1
8
1132
XM_017007629.1
XP_016863118.1
10
3126
XM_017007630.1
XP_016863119.1
8
1039
XM_017007631.1
XP_016863120.1
9
3084
XM_017007632.1
XP_016863121.1
7
978
XM_017007633.1
XP_016863122.1
9
3042
XM_017007634.1
XP_016863123.1
3
452
GenBank mRNA
AB020721.2
-
4
260
AK027138.1
-
4
383
AK091787.1
-
3
398
AK091881.1
-
11
1622
AK296932.1
-
2
328
BC086875.1
-
3
342
30708091
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0