Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005608.2 | NP_001005608.2 | 25 | 3396 | ||
NM_133663.2 | NP_598424.2 | 25 | 3396 | |||
GenBank mRNA | AK048888.1 | - | 25 | 3281 | ||
AK144111.1 | - | 2 | 392 | |||
AK145317.1 | - | 2 | 472 | |||
AK147424.1 | - | 25 | 3394 | |||
BC006632.1 | - | 5 | 442 | |||
BC059192.1 | - | 21 | 2591 | |||
L04678.1 | - | 26 | 3240 |