Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005905.1 | NP_001005905.1 | 4, 5 | 304 | ||
XM_006241371.2 | XP_006241433.1 | 4, 5 | 353 | |||
GenBank mRNA | BC083650.1 | - | 4, 5 | 304 | 7135109 | |
FQ223115.1 | - | 1, 2 | 25 |