Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159598.1 | NP_001153070.1 | 9 | 2675 | ||
NM_009733.2 | NP_033863.2 | 10 | 2783 | |||
GenBank mRNA | AA259709.1 | - | 1 | 41 | ||
AF009011.1 | - | 10 | 2735 | |||
BC011544.1 | - | 10 | 2502 | |||
BC055491.1 | - | 5 | 1062 | |||
BC069954.1 | - | 3 | 265 | |||
BC113171.1 | - | 9 | 2407 | 40061055 |