Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012535.2 | NP_036667.1 | 4 | 356 | ||
GenBank mRNA | BC097255.1 | - | 4 | 382 | 7444166 | |
M13749.1 | - | 4 | 356 |