Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015082.1 | NP_055897.1 | 3 | 363 | ||
XM_011543283.1 | XP_011541585.1 | 3 | 270 | |||
XM_011543284.1 | XP_011541586.1 | 3 | 341 | |||
GenBank mRNA | AK055684.1 | - | 3 | 341 | ||
BC024300.1 | - | 3 | 395 | 4662520 | ||
BX640845.1 | - | 3 | 285 |