Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243403.1 | NP_001230332.1 | 5 | 789 | ||
NM_015226.2 | NP_056041.1 | 5 | 789 | |||
XM_005255210.1 | XP_005255267.1 | 5 | 783 | |||
XM_005255211.1 | XP_005255268.1 | 5 | 783 | |||
XM_005255213.1 | XP_005255270.1 | 5 | 783 | |||
XM_005255214.1 | XP_005255271.1 | 5 | 783 | |||
XM_005255215.3 | XP_005255272.1 | 5 | 783 | |||
XM_005255216.1 | XP_005255273.1 | 5 | 783 | |||
XM_006720870.3 | XP_006720933.1 | 5 | 783 | |||
XM_011522434.1 | XP_011520736.1 | 5 | 783 | |||
XM_011522435.1 | XP_011520737.1 | 5 | 783 | |||
XM_011522436.2 | XP_011520738.1 | 5 | 783 | |||
XM_011522437.2 | XP_011520739.1 | 5 | 783 | |||
XM_011522438.2 | XP_011520740.1 | 5 | 783 | |||
XM_011522439.2 | XP_011520741.1 | 5 | 783 | |||
XM_011522440.2 | XP_011520742.1 | 5 | 783 | |||
XM_017023089.1 | XP_016878578.1 | 5 | 783 | |||
GenBank mRNA | AB002348.3 | - | 5 | 692 | ||
BC112897.1 | - | 5 | 696 | 4419248 |