Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 14 | 5435 | ||
NM_015349.2 | NP_056164.1 | 12 | 5168 | |||
GenBank mRNA | AK128771.1 | - | 1 | 1297 | ||
AK294706.1 | - | 13 | 2913 | |||
AL833540.1 | - | 12 | 5168 | |||
BC047630.1 | - | 1 | 2139 | |||
CR627378.1 | - | 13 | 5133 | |||
D87077.2 | - | 15 | 5332 |