Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015353.2 | NP_056168.1 | 6 | 2573 | ||
NR_110834.1 | 7 | 2454 | ||||
NR_110835.1 | 8 | 2536 | ||||
GenBank mRNA | AF055014.1 | - | 1 | 556 | ||
BC033329.1 | - | 7 | 2454 | |||
BC080561.1 | - | 1 | 1525 | |||
D79998.1 | - | 6 | 2512 |