Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027366.1 | NP_081642.1 | 3 | 713 | ||
GenBank mRNA | AK009303.1 | - | 3 | 570 |