Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033117.2 | NP_001028289.1 | 20 | 3154 | ||
NM_014850.3 | NP_055665.1 | 20 | 3226 | |||
XM_011534295.1 | XP_011532597.1 | 21 | 3241 | |||
XM_011534296.1 | XP_011532598.1 | 22 | 3358 | |||
XM_011534301.2 | XP_011532603.1 | 18 | 5070 | |||
XM_011534303.1 | XP_011532605.1 | 16 | 1966 | |||
XM_017007574.1 | XP_016863063.1 | 20 | 3564 | |||
XM_017007575.1 | XP_016863064.1 | 20 | 3492 | |||
XM_017007576.1 | XP_016863065.1 | 22 | 2733 | |||
XM_017007577.1 | XP_016863066.1 | 22 | 2672 | |||
XM_017007578.1 | XP_016863067.1 | 20 | 3592 | |||
XM_017007579.1 | XP_016863068.1 | 20 | 3560 | |||
XM_017007580.1 | XP_016863069.1 | 22 | 2721 | |||
GenBank mRNA | AB007871.2 | - | 20 | 2378 | ||
AF427144.1 | - | 20 | 2594 | |||
AF464189.1 | - | 20 | 2682 |