Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000267.1 | NP_001000267.1 | 1 | 260 |
Gene Symbol: LOC686288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101012.1 | NP_001094482.1 | 4 | 742 | ||
GenBank mRNA | BC128716.1 | - | 4 | 742 | 7128779 |