Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014878.4 | NP_055693.4 | 9, 10 | 1061 | ||
GenBank mRNA | AF490254.1 | - | 6, 7 | 545 | ||
AK225953.1 | - | 9, 10 | 1026 | |||
AK292169.1 | - | 9, 10 | 1004 | |||
AK315531.1 | - | 9, 10 | 1040 | |||
AL832239.1 | - | 9, 10 | 1027 | |||
AL832245.1 | - | 9, 10 | 1020 | |||
AY047588.1 | - | 2, 3 | 263 | |||
AY047589.1 | - | 2, 3 | 263 | |||
AY047590.1 | - | 2, 3 | 263 | |||
BC016137.2 | - | 8, 9 | 965 | 3908516 | ||
CR456957.1 | - | 6, 7 | 545 | |||
D13645.1 | - | 8, 9 | 963 |