Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 21 | 2417 | ||
NM_001316677.1 | NP_001303606.1 | 19 | 2090 | |||
NM_001323354.1 | NP_001310283.1 | 19 | 3810 | |||
NM_001323355.1 | NP_001310284.1 | 19 | 2223 | |||
NM_001323356.1 | NP_001310285.1 | 19 | 2217 | |||
NM_001323357.1 | NP_001310286.1 | 16 | 1802 | |||
NM_006504.5 | NP_006495.1 | 20 | 2246 | |||
NM_130435.4 | NP_569119.1 | 17 | 1893 | |||
XM_005252691.2 | XP_005252748.1 | 19 | 2090 | |||
XM_011539994.2 | XP_011538296.1 | 19 | 2205 | |||
XM_011539996.1 | XP_011538298.1 | 18 | 2082 | |||
XM_017016467.1 | XP_016871956.1 | 19 | 2208 | |||
XM_017016468.1 | XP_016871957.1 | 19 | 1997 | |||
GenBank mRNA | AJ315969.1 | - | 17 | 1802 | ||
AJ430580.1 | - | 17 | 2651 | |||
AK290414.1 | - | 17 | 1829 | |||
AK291828.1 | - | 19 | 2090 | |||
BX648180.1 | - | 14 | 4573 | |||
X54134.1 | - | 20 | 2018 |