Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 1 | 406 | ||
NM_014903.5 | NP_055718.4 | 1 | 406 | |||
XM_005269215.3 | XP_005269272.1 | 1 | 4407 | |||
XM_011538940.2 | XP_011537242.1 | 1 | 4408 | |||
XM_011538941.2 | XP_011537243.1 | 1 | 4407 | |||
XM_011538944.2 | XP_011537246.1 | 1 | 4405 | |||
XM_017020164.1 | XP_016875653.1 | 1 | 4409 | |||
XM_017020165.1 | XP_016875654.1 | 1 | 4409 | |||
XM_017020168.1 | XP_016875657.1 | 1 | 4408 | |||
XM_017020169.1 | XP_016875658.1 | 1 | 4407 | |||
XM_017020170.1 | XP_016875659.1 | 1 | 4404 | |||
GenBank mRNA | AJ488201.1 | - | 1 | 278 | ||
BC017667.2 | - | 1 | 783 | |||
DR005268.1 | - | 1 | 411 |